| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014432.hg.1 |
ENST00000262854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014432.hg.1 |
ENST00000342160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014432.hg.1 |
ENST00000474288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014432.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014432.hg.1 |
TC0X001067.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014429.hg.1 |
ENST00000262854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014429.hg.1 |
ENST00000342160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014429.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014429.hg.1 |
TC0X001067.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000262854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000342160.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000426907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000463852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000468322.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000612484.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000262854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000342160.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000426907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000463852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000468322.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
ENST00000612484.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166749.hg.1 |
TC0X00009767.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000262854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000342160.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000474288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000612484.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000262854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000342160.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000474288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
ENST00000612484.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166752.hg.1 |
TC0X00009767.hg.1 |
Clariom D TC |