| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366512.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000366513.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
ENST00000483271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_001139459.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012272.hg.1 |
NM_152609.2 |
Refseq |