Assay Details
| Gene Symbol: ZNF326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320185.1 | NP_001307114.1 | 4 - 5 | 364 | 77 | |
| GenBank mRNA | BC014899.1 | - | 4 - 5 | 322 | 77 | |
More Information
Set Membership: |
|

ZNF326
Genomic Map