Assay Details
| Gene Symbol: GCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006836.1 | NP_006827.1 | 31 - 32 | 3712 | 95 | |
| GenBank mRNA | BC153881.1 | - | 31 - 32 | 3712 | 8860118 | 95 |
| BU186578.1 | - | 2 - 3 | 171 | 95 | ||
| D86973.2 | - | 31 - 32 | 3712 | 95 | ||
| U77700.1 | - | 9 - 10 | 1177 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002034.hg.1
Set Membership: |
|

GCN1
Genomic Map