Assay Details
| Gene Symbol: GCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006836.1 | NP_006827.1 | 43 - 44 | 5629 | 80 | |
| GenBank mRNA | BC153881.1 | - | 43 - 44 | 5629 | 8860118 | 80 |
| D86973.2 | - | 43 - 44 | 5629 | 80 | ||
| U77700.1 | - | 21 - 22 | 3094 | 80 | ||
| U88836.1 | - | 4 - 5 | 587 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002034.hg.1
Set Membership: |
|

GCN1
Genomic Map