Assay Details
| Gene Symbol: RUBCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145642.3 | NP_001139114.1 | 6 - 7 | 1090 | 72 | |
| NM_014687.2 | NP_055502.1 | 6 - 7 | 931 | 72 | ||
| XM_005269374.3 | XP_005269431.1 | 6 - 7 | 926 | 72 | ||
| XM_006713827.3 | XP_006713890.1 | 6 - 7 | 927 | 72 | ||
| XM_006713828.3 | XP_006713891.1 | 6 - 7 | 926 | 72 | ||
| XM_006713829.3 | XP_006713892.1 | 6 - 7 | 1242 | 72 | ||
| XM_006713830.3 | XP_006713893.1 | 5 - 6 | 775 | 72 | ||
| XM_006713831.3 | XP_006713894.1 | 5 - 6 | 1088 | 72 | ||
| XM_017007543.1 | XP_016863032.1 | 5 - 6 | 771 | 72 | ||
| XM_017007544.1 | XP_016863033.1 | 5 - 6 | 772 | 72 | ||
| XM_017007545.1 | XP_016863034.1 | 5 - 6 | 1088 | 72 | ||
| GenBank mRNA | BC033615.1 | - | 6 - 7 | 907 | 72 | |
| BX647598.1 | - | 5 - 6 | 663 | 72 | ||
| D86979.3 | - | 6 - 7 | 779 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002182.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
phagocytosis
negative regulation of autophagy
negative regulation of phosphatidylinositol 3-kinase activity
negative regulation of endocytosis
multivesicular body sorting pathway
autophagosome maturation
negative regulation of autophagosome maturation
protein binding
immune system process
phagocytosis
negative regulation of autophagy
negative regulation of phosphatidylinositol 3-kinase activity
negative regulation of endocytosis
multivesicular body sorting pathway
autophagosome maturation
negative regulation of autophagosome maturation
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000273582.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000296343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000401223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000413360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000415452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000447048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000447048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000447048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000449205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000467303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000467303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000467303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000467303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000467303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000467303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000467303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000471364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000471364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000471364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000471364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000472149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000472149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000474214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000474214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000474214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000474214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | ENST00000474214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_001145642.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NM_014687.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013763.hg.1 | NR_030627.1 | Refseq |

RUBCN
Genomic Map