Assay Details
| Gene Symbol: TRGV9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC072396.1 | - | 2 - 3 | 426 | 72 | |
| M16768.1 | - | 1 - 2 | 201 | 72 | ||
| Gene Symbol: TARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | KC170727.1 | - | 1 - 2 | 52 | 72 | |
| M27335.1 | - | 1 - 2 | 49 | 72 | ||
| X72500.1 | - | 1 - 2 | 49 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |

TRGV9
Genomic Map