Assay Details
| Gene Symbol: IL23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ506993.1 | - | 2 - 3 | 335 | 53 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390400.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390400.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390400.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000390418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000633713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013479.hg.1 | ENST00000634000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390399.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390400.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390400.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390400.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000390418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000466254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610416.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610566.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000610847.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000612107.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000613761.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000616837.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000617717.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000633705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000633713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013479.hg.1 | ENST00000634000.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 185597 | - | - |

IL23A
Genomic Map