Assay Details
| Gene Symbol: EPHA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281765.2 | NP_001268694.1 | 9 - 10 | 2610 | 72 | |
| NM_001281767.2 | NP_001268696.1 | 9 - 10 | 2610 | 72 | ||
| NM_001318761.1 | NP_001305690.1 | 9 - 10 | 1728 | 72 | ||
| NM_004439.7 | NP_004430.4 | 9 - 10 | 2607 | 72 | ||
| XM_005265653.3 | XP_005265710.1 | 7 - 8 | 2461 | 72 | ||
| XM_011531735.2 | XP_011530037.1 | 8 - 9 | 2617 | 72 | ||
| GenBank mRNA | AB209375.1 | - | 9 - 10 | 2610 | 72 | |
| AK307826.1 | - | 9 - 10 | 2049 | 72 | ||
| BC136257.1 | - | 9 - 10 | 1877 | 9020244 | 72 | |
| BC136258.1 | - | 9 - 10 | 1877 | 9020245 | 72 | |
More Information
Set Membership: |
|

EPHA5
Genomic Map