Assay Details
| Gene Symbol: C1QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172369.3 | NP_758957.2 | 1 - 2 | 88 | 58 | |
| GenBank mRNA | AF087892.1 | - | 1 - 2 | 86 | 58 | |
| BC009016.1 | - | 1 - 2 | 53 | 4212772 | 58 | |
| DA849505.1 | - | 1 - 2 | 80 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF087892.1, BC009016.1, NM_172369.3 transcripts.
Set Membership: |
|

C1QC
Genomic Map