Assay Details
| Gene Symbol: NDUFAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250790.2 | XP_005250847.1 | 2 - 3 | 316 | 62 | |
| GenBank mRNA | BC028166.1 | - | 2 - 3 | 316 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250790.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000396124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000454358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000517976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518608.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518608.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518608.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518608.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000518608.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000519804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000519804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000519804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000519804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000520632.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000522683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000522683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000522683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000522683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000522683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000522683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | ENST00000523337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012331.hg.1 | NM_152416.3 | Refseq |

NDUFAF6
Genomic Map