Assay Details
| Gene Symbol: VSIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170553.1 | NP_001164024.1 | 4 - 5 | 680 | 72 | |
| NM_182607.4 | NP_872413.1 | 3 - 4 | 572 | 72 | ||
| XM_005262127.4 | XP_005262184.1 | 5 - 6 | 782 | 72 | ||
| XM_011530936.2 | XP_011529238.1 | 5 - 6 | 641 | 72 | ||
| GenBank mRNA | AK292472.1 | - | 3 - 4 | 451 | 72 | |
| AK301311.1 | - | 4 - 5 | 580 | 72 | ||
| AK315846.1 | - | 3 - 4 | 477 | 72 | ||
| BC043216.2 | - | 3 - 4 | 528 | 5295297 | 72 | |
| BX640913.1 | - | 3 - 4 | 486 | 72 | ||
| BX648658.1 | - | 3 - 4 | 572 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000529.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007079.hg.1 | ENST00000217957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007079.hg.1 | ENST00000415430.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007079.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007079.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007079.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007079.hg.1 | TC0X000529.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007075.hg.1 | ENST00000415430.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007075.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007075.hg.1 | TC0X000529.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000479635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000479635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000479635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155248.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155248.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155248.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155248.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155248.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155248.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155248.hg.1 | TC0X00008080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155252.hg.1 | TC0X00008080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000217957.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000415430.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000458383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000479635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000479635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000479635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | ENST00000485533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_001170553.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008080.hg.1 | NM_182607.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 147224 | - | - |

VSIG1
Genomic Map