| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110187.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012794.hg.1 |
NR_110187.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110187.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012794.hg.1 |
NR_110187.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195717.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195717.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195717.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195717.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195717.hg.1 |
TC0300012794.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300195718.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195718.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195718.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195718.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195718.hg.1 |
TC0300012794.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300195722.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195722.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195722.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195722.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195722.hg.1 |
TC0300012794.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300195725.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195725.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195725.hg.1 |
TC0300012794.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300195726.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195726.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195726.hg.1 |
TC0300012794.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300195719.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195719.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195719.hg.1 |
ENST00000471222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195719.hg.1 |
NR_110186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195719.hg.1 |
TC0300012794.hg.1 |
Clariom D TC |