Assay Details
| Gene Symbol: SPG20-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045180.1 | 5 - 6 | 529 | 69 | ||
| NR_045181.1 | 4 - 5 | 442 | 69 | |||
| GenBank mRNA | AW292824.1 | - | 5 - 6 | 528 | 69 | |
| BF732820.1 | - | 4 - 5 | 442 | 69 | ||
| CD674079.1 | - | 1 - 2 | 155 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000123.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13001394.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001394.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001394.hg.1 | TC13000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001392.hg.1 | ENST00000493739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001392.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001392.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001392.hg.1 | TC13000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001393.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001393.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001393.hg.1 | TC13000123.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146763.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300146763.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146763.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146763.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300146763.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146763.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146763.hg.1 | TC1300006877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | ENST00000488319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | ENST00000493739.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146762.hg.1 | TC1300006877.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300146765.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146765.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146765.hg.1 | NR_045180.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146765.hg.1 | NR_045181.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300146765.hg.1 | TC1300006877.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 213731 | - | - |

SPG20-AS1
Genomic Map