| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012705.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012705.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012705.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012705.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012705.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012705.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187628.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187628.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187628.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187628.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187628.hg.1 |
TC0600012705.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600012705.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012705.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012705.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012705.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012705.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012705.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187629.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187629.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187629.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187629.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187629.hg.1 |
TC0600012705.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600187630.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187630.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187630.hg.1 |
ENST00000369123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187630.hg.1 |
NR_046407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187630.hg.1 |
TC0600012705.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
244175_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
244175_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
244175_at9 |
- |
- |
| Human Genome U133 Set |
244175_PM_at |
- |
- |
| Human Genome U133 Set |
244175_at |
- |
- |