Assay Details
| Gene Symbol: C5orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294337.1 | NP_001281266.1 | 1 - 2 | 341 | 67 | |
| GenBank mRNA | BC171731.1 | - | 1 - 2 | 244 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145023.hg.1 | TC0500006558.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145021.hg.1 | TC0500006558.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |

C5orf38
Genomic Map