Assay Details
| Gene Symbol: SEMA6A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109879.1 | 1 - 2 | 296 | 89 | ||
| GenBank mRNA | CD300011.1 | - | 1 - 2 | 191 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157534.hg.1 | TC0500008402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157533.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157533.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157533.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157533.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157533.hg.1 | TC0500008402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500157532.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157532.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157532.hg.1 | TC0500008402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157537.hg.1 | TC0500008402.hg.1 | Clariom D TC |

SEMA6A-AS1
Genomic Map