| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003733.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003733.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003733.hg.1 |
TC22000200.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003734.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003734.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22003734.hg.1 |
TC22000200.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149155.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149155.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149155.hg.1 |
TC2200007032.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200149155.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149155.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149154.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149154.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149154.hg.1 |
TC2200007032.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200149154.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149154.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149156.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149156.hg.1 |
NM_182527.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200149156.hg.1 |
TC2200007032.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200149156.hg.1 |
ENST00000216144.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200149156.hg.1 |
NM_182527.2 |
Refseq |