Assay Details
| Gene Symbol: DHRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024308.3 | NP_077284.2 | 1 - 2 | 357 | 61 | |
| XM_005257658.2 | XP_005257715.1 | 1 - 2 | 362 | 61 | ||
| GenBank mRNA | AK315735.1 | - | 1 - 2 | 278 | 61 | |
| AY358712.1 | - | 1 - 2 | 254 | 61 | ||
| BC002731.2 | - | 1 - 2 | 243 | 3631177 | 61 | |
| BG037061.1 | - | 1 - 2 | 360 | 61 | ||
| LC110386.1 | - | 1 - 2 | 143 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000439.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK315735.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17005223.hg.1 | ENST00000251312.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005223.hg.1 | ENST00000394445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005223.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005223.hg.1 | TC17000439.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005218.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005218.hg.1 | TC17000439.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005221.hg.1 | ENST00000251312.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005221.hg.1 | ENST00000394445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005221.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005221.hg.1 | TC17000439.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155103.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155103.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155103.hg.1 | TC1700007641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700155103.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155103.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | TC1700007641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155107.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | TC1700007641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155112.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | TC1700007641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155113.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | TC1700007641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155109.hg.1 | NM_024308.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 206021 | - | - |

DHRS11
Genomic Map