Assay Details
| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI160094.1 | - | 1 - 2 | 395 | 69 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000308961.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000418389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000591341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000592634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | ENST00000593233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_001193375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NM_175614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011851.hg.1 | NR_034166.2 | Refseq |

NDUFA11
Genomic Map