| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023618.hg.1 |
ENST00000310442.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023618.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023618.hg.1 |
ENST00000526986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023618.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023618.hg.1 |
TC11001969.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023617.hg.1 |
ENST00000310442.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023617.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023617.hg.1 |
ENST00000526986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023617.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023617.hg.1 |
TC11001969.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
ENST00000602427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023620.hg.1 |
ENST00000310442.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023620.hg.1 |
ENST00000526986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023620.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023620.hg.1 |
TC11001969.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100186925.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186925.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186925.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186925.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186925.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186925.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186925.hg.1 |
TC1100011291.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186921.hg.1 |
TC1100011291.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186920.hg.1 |
TC1100011291.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000310442.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000525925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000526986.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000534073.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
ENST00000602427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011291.hg.1 |
NM_207340.2 |
Refseq |