| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007701.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153676.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153676.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153676.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153676.hg.1 |
TC2000007701.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153676.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153676.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153676.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153677.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153677.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153677.hg.1 |
TC2000007701.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153677.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153677.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
ENST00000435177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
ENST00000452336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
NR_111906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007701.hg.1 |
NR_111906.1 |
Refseq |