Assay Details
| Gene Symbol: ZNF704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005251279.4 | XP_005251336.2 | 1 - 2 | 842 | 105 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000327835.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000517986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000519936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000520336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | ENST00000522040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010918.hg.1 | NM_001033723.2 | Refseq |

ZNF704
Genomic Map