| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000291458.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000460810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000466854.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000479899.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000497726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
ENST00000498466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001017928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012186.hg.1 |
NM_001308326.1 |
Refseq |