Assay Details
| Gene Symbol: CCDC58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017928.3 | NP_001017928.1 | 4 - 5 | 389 | 109 | |
| NM_001308326.1 | NP_001295255.1 | 4 - 5 | 415 | 109 | ||
| XM_011512410.2 | XP_011510712.1 | 4 - 5 | 367 | 109 | ||
| GenBank mRNA | BC054051.1 | - | 4 - 5 | 380 | 109 | |
| BC062725.1 | - | 4 - 5 | 389 | 6495960 | 109 | |
| BC109378.1 | - | 4 - 5 | 373 | 109 | ||
| CD683826.1 | - | 4 - 5 | 411 | 109 | ||
| DA025111.1 | - | 4 - 5 | 411 | 109 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000592203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000592203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000291458.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000460810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000466854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000479899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000497726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | ENST00000498466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001017928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012186.hg.1 | NM_001308326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000586882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000588744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000592203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000592203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | ENST00000592801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009217.hg.1 | NR_049896.1 | Refseq |

CCDC58
Genomic Map