| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014981.hg.1 |
ENST00000281441.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014981.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014981.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014981.hg.1 |
TC11001192.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014982.hg.1 |
ENST00000281441.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014982.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014982.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014982.hg.1 |
TC11001192.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014975.hg.1 |
ENST00000281441.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014975.hg.1 |
ENST00000525709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014975.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014975.hg.1 |
TC11001192.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014980.hg.1 |
ENST00000281441.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014980.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014980.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014980.hg.1 |
TC11001192.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100171492.hg.1 |
TC1100009514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171493.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171493.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171493.hg.1 |
TC1100009514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171497.hg.1 |
TC1100009514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171489.hg.1 |
TC1100009514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171495.hg.1 |
TC1100009514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171498.hg.1 |
TC1100009514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000281441.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000524567.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000525709.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000527754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
ENST00000529381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009514.hg.1 |
NM_138788.4 |
Refseq |