Assay Details
| Gene Symbol: ARHGAP35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004491.4 | NP_004482.4 | 1 - 2 | 3681 | 72 | |
| XM_011526873.2 | XP_011525175.1 | 2 - 3 | 4077 | 72 | ||
| XM_017026714.1 | XP_016882203.1 | 2 - 3 | 4077 | 72 | ||
| GenBank mRNA | AB051509.2 | - | 1 - 2 | 3681 | 72 | |
| AF159851.1 | - | 1 - 2 | 3684 | 72 | ||
| AK298356.1 | - | 1 - 2 | 505 | 72 | ||
| BC150257.1 | - | 1 - 2 | 3689 | 9007187 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000669.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neural tube closure
transcription, DNA-templated
small GTPase mediated signal transduction
regulation of actin polymerization or depolymerization
regulation of cell shape
positive regulation of neuron projection development
forebrain development
negative regulation of Rho protein signal transduction
camera-type eye development
negative regulation of vascular permeability
positive regulation of GTPase activity
positive regulation of cilium assembly
negative regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription corepressor activity
GTPase activator activity
GTP binding
negative regulation of transcription from RNA polymerase II promoter
neural tube closure
transcription, DNA-templated
small GTPase mediated signal transduction
regulation of actin polymerization or depolymerization
regulation of cell shape
positive regulation of neuron projection development
forebrain development
negative regulation of Rho protein signal transduction
camera-type eye development
negative regulation of vascular permeability
positive regulation of GTPase activity
positive regulation of cilium assembly
negative regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription corepressor activity
GTPase activator activity
GTP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19009365.hg.1 | ENST00000317082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009365.hg.1 | ENST00000404338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009365.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009365.hg.1 | TC19000669.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000596593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000596593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000598548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000598548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000598548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009363.hg.1 | ENST00000317082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009363.hg.1 | ENST00000404338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009363.hg.1 | ENST00000501576.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009363.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009363.hg.1 | TC19000669.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009364.hg.1 | ENST00000317082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009364.hg.1 | ENST00000404338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009364.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009364.hg.1 | TC19000669.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | TC1900008399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164514.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | TC1900008399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164511.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000595822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000596593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000596593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000598548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000598548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000598548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000599284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | ENST00000615647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008399.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164512.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164512.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164512.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164512.hg.1 | TC1900008399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164512.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164512.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164512.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164513.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164513.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164513.hg.1 | NM_004491.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164513.hg.1 | TC1900008399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164513.hg.1 | ENST00000404338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164513.hg.1 | ENST00000614079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164513.hg.1 | NM_004491.4 | Refseq |

ARHGAP35
Genomic Map