| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031556.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031556.hg.1 |
ENST00000497869.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031556.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031556.hg.1 |
TC01002086.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031559.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031559.hg.1 |
TC01002086.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031557.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031557.hg.1 |
ENST00000497869.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031557.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031557.hg.1 |
TC01002086.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100191493.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191493.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191493.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191493.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191493.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191493.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191493.hg.1 |
TC0100012453.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000453580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012453.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191497.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191497.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191497.hg.1 |
TC0100012453.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100191494.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191494.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191494.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191494.hg.1 |
ENST00000379236.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191494.hg.1 |
ENST00000497869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191494.hg.1 |
NM_003327.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191494.hg.1 |
TC0100012453.hg.1 |
Clariom D TC |