Assay Details
| Gene Symbol: RHPN1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026785.1 | 1 | 840 | 77 | ||
| GenBank mRNA | AK026503.1 | - | 1 | 751 | 77 | |
| BC000203.1 | - | 1 | 753 | 77 | ||
| BI829214.1 | - | 1 | 517 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001714.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 195380 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 273456 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P71503 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P71503 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 40286 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42024 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P16709593 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P71503 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P71503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532328 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16146286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817039 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817019 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16146282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16146287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532322 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532322 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817019 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16146287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817039 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16146286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26817030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16146282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06532328 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P71503 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P122700 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P71503 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 1483 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 3567 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16709593 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32363438 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532327 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532282 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532308 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532306 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16146285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532298 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16146287 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532275 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16146274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532279 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532290 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532322 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532281 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532287 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16146288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532296 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16146277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16146282 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532293 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532318 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532325 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532311 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06532332 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P16709593 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8153408 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8153407 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8153406 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8153405 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 207458_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022604.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022604.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022604.hg.1 | TC08001714.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022605.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022605.hg.1 | TC08001714.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | ENST00000596598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182584.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182584.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182584.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182584.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182584.hg.1 | TC0800012128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800182585.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182585.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182585.hg.1 | TC0800012128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800182583.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182583.hg.1 | ENST00000596598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182583.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182583.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182583.hg.1 | ENST00000596598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182583.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182583.hg.1 | TC0800012128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | ENST00000596598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012128.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182582.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182582.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182582.hg.1 | ENST00000518049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182582.hg.1 | NR_026785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182582.hg.1 | TC0800012128.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 207458_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207458_at | - | - |
| Human Genome U133A 2.0 Array | 207458_at | - | - |
| Human Genome U133A 2.0 Array | 207458_PM_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw597873 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207458_at11 | - | - |
| Human Genome U133 Set | 207458_PM_at | - | - |
| Human Genome U133 Set | 207458_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036214 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 137078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1688772 | BM685262.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1688772 | BC000203.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1688772 | BM968345.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1688772 | NR_026785.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1688772 | AK026503.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1688772 | BM685262.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1688772 | BC000203.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1688772 | BM968345.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1688772 | NR_026785.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1688772 | AK026503.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1688772 | BM685262.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1688772 | BC000203.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1688772 | BM968345.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1688772 | NR_026785.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1688772 | AK026503.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1688772 | BM685262.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1688772 | BC000203.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1688772 | BM968345.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1688772 | NR_026785.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1688772 | AK026503.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1688772 | BM685262.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1688772 | BC000203.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1688772 | BM968345.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1688772 | NR_026785.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1688772 | AK026503.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1688772 | BM685262.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1688772 | BC000203.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1688772 | BM968345.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1688772 | NR_026785.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1688772 | AK026503.1 | human_transcriptome |

RHPN1-AS1
Genomic Map