| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001460.hg.1 |
ENST00000327473.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001460.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001460.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001460.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001460.hg.1 |
TC19000091.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001457.hg.1 |
ENST00000327473.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001457.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001457.hg.1 |
TC19000091.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
TC1900006711.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900147110.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900147105.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147105.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147105.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900147105.hg.1 |
TC1900006711.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900147105.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147105.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900147105.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000327473.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000536716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
ENST00000598107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_001167942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006711.hg.1 |
NM_152362.2 |
Refseq |