Assay Details
| Gene Symbol: APCDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153000.4 | NP_694545.1 | 2 - 3 | 600 | 88 | |
| XM_011525603.1 | XP_011523905.1 | 2 - 3 | 593 | 88 | ||
| GenBank mRNA | AB056722.1 | - | 2 - 3 | 599 | 88 | |
| AB104887.1 | - | 2 - 3 | 599 | 88 | ||
| AF461902.1 | - | 1 - 2 | 197 | 88 | ||
| AK300743.1 | - | 3 - 4 | 402 | 88 | ||
| AK303076.1 | - | 1 - 2 | 192 | 88 | ||
| BC053324.1 | - | 3 - 4 | 571 | 6644230 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000051.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF461902.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000558.hg.1 | ENST00000355285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000558.hg.1 | ENST00000423585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000558.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000558.hg.1 | TC18000051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145645.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145646.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145646.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145646.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145646.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145646.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145646.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145646.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |

APCDD1
Genomic Map