Assay Details
| Gene Symbol: SPRY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030964.3 | NP_112226.2 | 2 - 3 | 266 | 73 | |
| XM_011537685.2 | XP_011535987.1 | 2 - 3 | 993 | 73 | ||
| GenBank mRNA | AF227516.1 | - | 2 - 3 | 212 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001880.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF227516.1,NM_030964.3,XM_011537685.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05025995.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025995.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025995.hg.1 | TC05001880.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185711.hg.1 | TC0500012312.hg.1 | Clariom D TC |

SPRY4
Genomic Map