Assay Details
| Gene Symbol: ZNF785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK056692.1 | - | 2 - 3 | 450 | 109 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000395216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000470110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000562128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | ENST00000567773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009978.hg.1 | NM_152458.6 | Refseq |

ZNF785
Genomic Map