Assay Details
| Gene Symbol: LINC00889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026935.1 | 1 | 2007 | 129 | ||
| GenBank mRNA | AK055234.1 | - | 1 | 1556 | 129 | |
| AK055382.1 | - | 1 | 1691 | 129 | ||
| BC105719.1 | - | 1 | 1681 | 129 | ||
| BC105725.1 | - | 1 | 1681 | 129 | ||
| BC105726.1 | - | 1 | 1681 | 129 | ||
| BC105727.1 | - | 1 | 1681 | 129 | ||
| BC139831.1 | - | 1 | 1684 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001424.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32716978 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32716972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32716979 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32716974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32716977 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32716975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32716976 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 93526 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 24241 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P360268 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P360268 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 35031 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P137194 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P137194 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20934 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P137194 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P360268 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P360268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716978 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17403315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716981 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716984 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17403315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716978 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716981 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716984 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32716980 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P360268 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P137194 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P137194 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P137194 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P360268 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 131541 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 65218 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P137194 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17403314 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983723 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983737 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11983729 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P137194 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8175443 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8175442 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553357_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018880.hg.1 | NR_026935.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018880.hg.1 | TC0X001424.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008741.hg.1 | ENST00000547679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174432.hg.1 | ENST00000315930.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174432.hg.1 | NR_026935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174432.hg.1 | ENST00000315930.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174432.hg.1 | NR_026935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174432.hg.1 | TC0X00010954.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1553357_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553357_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553357_at9 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040467 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040467 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 223468 | - | - |

LINC00889
Genomic Map