| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03032909.hg.1 |
ENST00000327532.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03032909.hg.1 |
ENST00000469243.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03032909.hg.1 |
ENST00000488595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03032909.hg.1 |
ENST00000536706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03032909.hg.1 |
NM_001170538.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03032909.hg.1 |
NM_173543.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03032909.hg.1 |
TC03001819.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000327532.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000466301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000469243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000488595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
NM_001170538.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
NM_173543.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000327532.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000466301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000469243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
ENST00000488595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
NM_001170538.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
NM_173543.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300193898.hg.1 |
TC0300012564.hg.1 |
Clariom D TC |