Assay Details
| Gene Symbol: PRR14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173566.2 | NP_775837.2 | 4 - 5 | 5952 | 67 | |
| XM_006724219.3 | XP_006724282.1 | 4 - 5 | 6512 | 67 | ||
| XM_006724220.3 | XP_006724283.1 | 4 - 5 | 5914 | 67 | ||
| XM_006724221.2 | XP_006724284.1 | 3 - 4 | 5468 | 67 | ||
| XM_011530080.2 | XP_011528382.1 | 5 - 6 | 6720 | 67 | ||
| XM_011530081.2 | XP_011528383.1 | 4 - 5 | 6596 | 67 | ||
| XM_017028736.1 | XP_016884225.1 | 4 - 5 | 7672 | 67 | ||
| XM_017028737.1 | XP_016884226.1 | 3 - 4 | 7110 | 67 | ||
| GenBank mRNA | AK130944.1 | - | 1 - 2 | 1766 | 67 | |
| BC040859.1 | - | 1 - 2 | 666 | 5535580 | 67 | |
| BX647272.1 | - | 1 - 2 | 255 | 67 | ||
| CT841517.1 | - | 3 - 4 | 5784 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001490.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22011936.hg.1 | ENST00000327423.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011936.hg.1 | ENST00000397493.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011936.hg.1 | ENST00000434485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011936.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011936.hg.1 | TC22001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011937.hg.1 | ENST00000327423.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011937.hg.1 | ENST00000397493.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011937.hg.1 | ENST00000434485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011937.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011937.hg.1 | TC22001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011938.hg.1 | ENST00000327423.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011938.hg.1 | ENST00000397493.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011938.hg.1 | ENST00000434485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011938.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011938.hg.1 | TC22001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011939.hg.1 | ENST00000327423.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011939.hg.1 | ENST00000397493.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011939.hg.1 | ENST00000434485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011939.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011939.hg.1 | TC22001490.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | TC2200008489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161153.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | TC2200008489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161154.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | TC2200008489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161151.hg.1 | NM_173566.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 222377 | - | - |

PRR14L
Genomic Map