| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005580.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005580.hg.1 |
ENST00000439993.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005580.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11005580.hg.1 |
TC11000461.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000533436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007670.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
ENST00000263314.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
ENST00000534820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
ENST00000616487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
NM_002559.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100153944.hg.1 |
TC1100007670.hg.1 |
Clariom D TC |