Assay Details
| Gene Symbol: BPIFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174897.2 | NP_777557.1 | 3 - 4 | 304 | 83 | |
| XM_017027663.1 | XP_016883152.1 | 3 - 4 | 352 | 83 | ||
| GenBank mRNA | AF465767.1 | - | 3 - 4 | 304 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000221.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20002748.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002748.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002748.hg.1 | TC20000221.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148871.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148871.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148871.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148871.hg.1 | TC2000007139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000148871.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148871.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148871.hg.1 | NM_174897.2 | Refseq |

BPIFB6
Genomic Map