Assay Details
| Gene Symbol: UHMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184763.1 | NP_001171692.1 | 5 - 6 | 782 | 63 | |
| NM_144624.2 | NP_653225.2 | 5 - 6 | 1123 | 63 | ||
| NM_175866.4 | NP_787062.1 | 5 - 6 | 1123 | 63 | ||
| GenBank mRNA | AJ536197.1 | - | 5 - 6 | 1072 | 63 | |
| AK058195.1 | - | 5 - 6 | 782 | 63 | ||
| AK292369.1 | - | 5 - 6 | 946 | 63 | ||
| AL834136.1 | - | 4 - 5 | 1508 | 63 | ||
| BC014917.1 | - | 4 - 5 | 612 | 63 | ||
| BC026046.1 | - | 5 - 6 | 1085 | 4649261 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001421.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
regulation of alternative mRNA splicing, via spliceosome
cell cycle arrest
peptidyl-serine phosphorylation
neuron projection development
positive regulation of translational initiation
protein autophosphorylation
regulation of protein export from nucleus
mRNA binding
protein serine/threonine kinase activity
protein binding
ATP binding
transferase activity
ribonucleoprotein complex binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
regulation of alternative mRNA splicing, via spliceosome
cell cycle arrest
peptidyl-serine phosphorylation
neuron projection development
positive regulation of translational initiation
protein autophosphorylation
regulation of protein export from nucleus
mRNA binding
protein serine/threonine kinase activity
protein binding
ATP binding
transferase activity
ribonucleoprotein complex binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | ENST00000538489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | ENST00000545294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022200.hg.1 | TC01001421.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010379.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000282169.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000489294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000538489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | ENST00000545294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | NM_001184763.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | NM_144624.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | NM_175866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176474.hg.1 | TC0100010379.hg.1 | Clariom D TC |

UHMK1
Genomic Map