Assay Details
| Gene Symbol: TRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317804.1 | NP_001304733.1 | 4 - 5 | 487 | 127 | |
| NM_001317805.1 | NP_001304734.1 | 3 - 4 | 1002 | 127 | ||
| NM_014294.5 | NP_055109.1 | 3 - 4 | 570 | 127 | ||
| GenBank mRNA | AK292777.1 | - | 3 - 4 | 354 | 127 | |
| AK303411.1 | - | 4 - 5 | 415 | 127 | ||
| BC000687.2 | - | 3 - 4 | 392 | 3347823 | 127 | |
| BC020283.1 | - | 3 - 4 | 385 | 127 | ||
| BC037738.1 | - | 3 - 4 | 482 | 5261490 | 127 | |
| BT007359.1 | - | 3 - 4 | 310 | 127 | ||
| CR541654.1 | - | 3 - 4 | 310 | 127 | ||
| CR541739.1 | - | 3 - 4 | 310 | 127 | ||
| X63679.1 | - | 3 - 4 | 431 | 127 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001312.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001317805.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08016904.hg.1 | ENST00000262213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016904.hg.1 | ENST00000521049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016904.hg.1 | ENST00000521425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016904.hg.1 | ENST00000536748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016904.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016904.hg.1 | TC08001312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016902.hg.1 | ENST00000262213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016902.hg.1 | ENST00000521425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016902.hg.1 | ENST00000536748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016902.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016902.hg.1 | TC08001312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173071.hg.1 | TC0800010765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173069.hg.1 | TC0800010765.hg.1 | Clariom D TC |

TRAM1
Genomic Map