Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022039.3 | NP_071322.1 | 1 - 2 | 609 | 74 | |
| NR_136613.1 | 1 - 2 | 609 | 74 | |||
| XM_005270053.2 | XP_005270110.2 | 1 - 2 | 660 | 74 | ||
| XM_017016570.1 | XP_016872059.1 | 1 - 2 | 660 | 74 | ||
| GenBank mRNA | AF281859.1 | - | 1 - 2 | 882 | 74 | |
| AK314735.1 | - | 1 - 2 | 554 | 74 | ||
| BC063415.1 | - | 1 - 2 | 376 | 5223794 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001609.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10020869.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020869.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020869.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020869.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020870.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020870.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020870.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020870.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020867.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020867.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020867.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020867.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020871.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020871.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020871.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020871.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020861.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020861.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020861.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020861.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020862.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020862.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020862.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020862.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020868.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020868.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020868.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020868.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020865.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020865.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020865.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020865.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000179657.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179657.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179657.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179657.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179657.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179657.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179657.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179658.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179660.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179665.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179665.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179665.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179665.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179665.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179666.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179666.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179666.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179666.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179666.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179664.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179664.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179664.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179664.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179664.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179662.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179662.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179662.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179662.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179662.hg.1 | TC1000011652.hg.1 | Clariom D TC |

FBXW4
Genomic Map