Assay Details
| Gene Symbol: WDR77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317062.1 | NP_001303991.1 | 3 - 4 | 586 | 67 | |
| NM_001317063.1 | NP_001303992.1 | 3 - 4 | 586 | 67 | ||
| NM_024102.3 | NP_077007.1 | 3 - 4 | 586 | 67 | ||
| GenBank mRNA | AB073603.1 | - | 3 - 4 | 534 | 67 | |
| AF052183.1 | - | 3 - 4 | 472 | 67 | ||
| AF478464.1 | - | 3 - 4 | 447 | 67 | ||
| AK022860.1 | - | 3 - 4 | 516 | 67 | ||
| AK223189.1 | - | 3 - 4 | 516 | 67 | ||
| AL557974.3 | - | 3 - 4 | 502 | 67 | ||
| AY225316.1 | - | 3 - 4 | 472 | 67 | ||
| BC001679.2 | - | 3 - 4 | 507 | 2822173 | 67 | |
| BC006477.1 | - | 3 - 4 | 507 | 2822173 | 67 | |
| BC009411.1 | - | 3 - 4 | 519 | 3614223 | 67 | |
| BC011778.2 | - | 3 - 4 | 526 | 3941878 | 67 | |
| BC016946.1 | - | 3 - 4 | 486 | 3845908 | 67 | |
| BI195029.1 | - | 2 - 3 | 258 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002985.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal snRNP assembly
regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
peptidyl-arginine N-methylation
secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development
negative regulation of epithelial cell proliferation involved in prostate gland development
protein binding
methyl-CpG binding
protein-arginine N-methyltransferase activity
ligand-dependent nuclear receptor transcription coactivator activity
spliceosomal snRNP assembly
regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
peptidyl-arginine N-methylation
secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development
negative regulation of epithelial cell proliferation involved in prostate gland development
protein binding
methyl-CpG binding
protein-arginine N-methyltransferase activity
ligand-dependent nuclear receptor transcription coactivator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01046355.hg.1 | ENST00000235090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046355.hg.1 | ENST00000411751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046355.hg.1 | ENST00000497278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046355.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046355.hg.1 | TC01002985.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046356.hg.1 | ENST00000235090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046356.hg.1 | ENST00000411751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046356.hg.1 | ENST00000497278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046356.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046356.hg.1 | TC01002985.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000416099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000416099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000416099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000416099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000416099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000416099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000445680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015273.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000235090.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000449340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000459665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | ENST00000497278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | NM_024102.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213886.hg.1 | TC0100015273.hg.1 | Clariom D TC |

WDR77
Genomic Map