| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000416099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000416099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000416099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046347.hg.1 |
ENST00000235090.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046347.hg.1 |
ENST00000411751.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046347.hg.1 |
ENST00000497278.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046347.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046347.hg.1 |
TC01002985.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213879.hg.1 |
TC0100015273.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000235090.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000416099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000416099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000416099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000445680.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000449340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000459665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
ENST00000497278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015273.hg.1 |
NM_024102.3 |
Refseq |