Assay Details
| Gene Symbol: CCDC50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174908.3 | NP_777568.1 | 5 - 6 | 1041 | 86 | |
| GenBank mRNA | AJ416916.2 | - | 5 - 6 | 1041 | 86 | |
| BC065004.1 | - | 5 - 6 | 538 | 6049272 | 86 | |
More Information
Set Membership: |
|

CCDC50
Genomic Map