Assay Details
| Gene Symbol: ICA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB053316.1 | - | 3 - 4 | 371 | 123 | |
| DB097704.1 | - | 3 - 4 | 231 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB053316.1 transcripts.
Set Membership: |
|

ICA1L
Genomic Map