Assay Details
| Gene Symbol: ADIPOQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177800.1 | NP_001171271.1 | 3 - 4 | 349 | 71 | |
| NM_004797.3 | NP_004788.1 | 2 - 3 | 298 | 71 | ||
| GenBank mRNA | AK291525.1 | - | 2 - 3 | 271 | 71 | |
| AK312868.1 | - | 3 - 4 | 319 | 71 | ||
| AX767959.1 | - | 2 - 3 | 240 | 71 | ||
| BC054496.1 | - | 239 | 6192794 | 71 | ||
| BC096308.3 | - | 2 - 3 | 282 | 40007540 | 71 | |
| BC096309.3 | - | 2 - 3 | 282 | 40007541 | 71 | |
| BC096310.3 | - | 2 - 3 | 282 | 40007542 | 71 | |
| BC096311.3 | - | 2 - 3 | 282 | 40007544 | 71 | |
| D45371.1 | - | 2 - 3 | 240 | 71 | ||
| EU420013.1 | - | 1 - 2 | 214 | 71 | ||
More Information
Set Membership: |
|

ADIPOQ
Genomic Map