Assay Details
| Gene Symbol: GPR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080865.3 | NP_543141.3 | 1 | 2117 | 126 | |
| GenBank mRNA | BC026357.1 | - | 1 | 2050 | 4812283 | 126 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237120 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22371924 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237122 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02237118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15438224 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 109852 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 18344 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P434289 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P434289 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36142 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P138167 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P138167 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12781 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P434289 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P434289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22371924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22371903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22371922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22371922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22371924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22371903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237127 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P434289 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P138167 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237107 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237113 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237130 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237111 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237115 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237122 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237116 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237129 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237124 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237110 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237126 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237105 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237118 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02237132 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P02237127 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438232 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P02237125 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438228 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P02237126 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438233 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438229 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438234 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438230 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438231 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438226 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15438227 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P138167 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P138167 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P434289 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 97710 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 35049 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P138167 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237073 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237185 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237130 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237155 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237181 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237191 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237099 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438241 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237082 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237107 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237201 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237105 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237111 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237089 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237115 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237087 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237164 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237097 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237110 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237085 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237124 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237079 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237183 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237161 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237126 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237129 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438234 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237159 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237094 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237209 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237176 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237174 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237118 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237187 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237169 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237198 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237157 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237208 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237076 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237203 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237113 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237179 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237206 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237100 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237096 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237091 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237211 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P138167 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_080865_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8080161 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8080159 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8080160 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8080158 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1554559_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11740505_at | - | - |
| Affymetrix Human Genome U219 Array | 11740505_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1554559_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1554559_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_080865_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554559_at4 | - | - |
| Human Genome U95 Set | 74224_at | - | - |
| Human Genome U95 Set | 74224_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022664 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022664 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 122459 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34878895-S | NM_080865.3 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34878895-S | BC026357.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_5373-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006400292 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006400292 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_5373 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_5373 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1756807 | BC026357.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1756807 | NM_080865.3 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1756807 | BC026357.1 | human_transcriptome |

GPR62
Genomic Map