Assay Details
| Gene Symbol: GOLGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002077.3 | NP_002068.1 | 14 - 15 | 1656 | 63 | |
| XM_005251929.3 | XP_005251986.1 | 14 - 15 | 1588 | 63 | ||
| XM_006717062.3 | XP_006717125.1 | 14 - 15 | 1652 | 63 | ||
| XM_006717063.3 | XP_006717126.1 | 14 - 15 | 1623 | 63 | ||
| GenBank mRNA | AB208858.1 | - | 9 - 10 | 1013 | 63 | |
| BC032853.1 | - | 14 - 15 | 1626 | 4828793 | 63 | |
| U51587.1 | - | 14 - 15 | 1648 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001583.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09019876.hg.1 | ENST00000373555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019876.hg.1 | ENST00000475407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019876.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019876.hg.1 | TC09001583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019875.hg.1 | ENST00000373555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019875.hg.1 | ENST00000475407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019875.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019875.hg.1 | TC09001583.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186638.hg.1 | TC0900012275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186640.hg.1 | TC0900012275.hg.1 | Clariom D TC |

GOLGA1
Genomic Map