Assay Details
| Gene Symbol: GOLGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002077.3 | NP_002068.1 | 2 - 3 | 185 | 80 | |
| XM_005251929.3 | XP_005251986.1 | 2 - 3 | 117 | 80 | ||
| XM_006717062.3 | XP_006717125.1 | 2 - 3 | 181 | 80 | ||
| XM_006717063.3 | XP_006717126.1 | 2 - 3 | 152 | 80 | ||
| GenBank mRNA | AK302445.1 | - | 2 - 3 | 115 | 80 | |
| BC032853.1 | - | 2 - 3 | 155 | 4828793 | 80 | |
| BG699932.1 | - | 2 - 3 | 158 | 80 | ||
| DA324844.1 | - | 2 - 3 | 186 | 80 | ||
| U51587.1 | - | 2 - 3 | 177 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001583.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK302445.1,BC032853.1,NM_002077.3,U51587.1,XM_005251929.3,XM_006717062.3,XM_006717063.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019895.hg.1 | ENST00000373555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019895.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019895.hg.1 | TC09001583.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900186660.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186660.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186660.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186660.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186660.hg.1 | TC0900012275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373551.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000475407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | ENST00000485337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012275.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186659.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186659.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186659.hg.1 | ENST00000373555.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186659.hg.1 | NM_002077.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186659.hg.1 | TC0900012275.hg.1 | Clariom D TC |

GOLGA1
Genomic Map