Assay Details
| Gene Symbol: GGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003878.2 | NP_003869.1 | 1 - 2 | 388 | 148 | |
| XM_011517623.2 | XP_011515925.1 | 1 - 2 | 139 | 148 | ||
| GenBank mRNA | AK290450.1 | - | 1 - 2 | 132 | 148 | |
| AK301094.1 | - | 1 - 2 | 138 | 148 | ||
| BC025025.1 | - | 1 - 2 | 189 | 3924620 | 148 | |
| U55206.1 | - | 1 - 2 | 164 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001269.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016364.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016364.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016364.hg.1 | TC08001269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016365.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016365.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016365.hg.1 | TC08001269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016368.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016368.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016368.hg.1 | TC08001269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016366.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016366.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016366.hg.1 | TC08001269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016370.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016370.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016370.hg.1 | TC08001269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016369.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016369.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016369.hg.1 | TC08001269.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800172187.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172187.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172187.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172187.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172187.hg.1 | TC0800010630.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172184.hg.1 | TC0800010630.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172183.hg.1 | TC0800010630.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172185.hg.1 | TC0800010630.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800172188.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172188.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172188.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172188.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172188.hg.1 | TC0800010630.hg.1 | Clariom D TC |

GGH
Genomic Map